VWR® Bossheads, Universal, Pivoting
Supplier: Avantor
VWR®
241-0113EA 129 EUR
241-0113
VWR® Bossheads, Universal, Pivoting
Clamps Bossheads
Bosshead made of laboral aluminium alloy for connecting two retort rods at any angle.
  • Tommy screw for securing the bosshead at the desired angle
  • T-screws for securing the retort rods