VWR® Bossheads
Supplier: VWR Collection
241-0119EA 913 EUR
241-0119 241-0120
VWR® Bossheads
Clamps Bossheads
For assembling 12, 13 and 16 mm diameter rods.
  • Bossheads can be fully opened; this enables the rod to be securely attached in the desired position
  • Easy to assemble and take apart by means of an Allen key (optional, 241-0053)
  • Excellent chemical resistance, robust
* When stainless steel boss is locked onto the stainless steel rod it cannot be moved by a rotational force less than 3,5 Nm.