VWR® Bossheads
Supplier: VWR Collection
241-0131EA 143 EUR
241-0131 241-0128 241-0224 241-0129 241-0130 241-0133
VWR® Bossheads
Clamps Bossheads
For assembling 8, 12, 13, 16 and 21 mm diameter rods.
  • Light, robust design
  • Bossheads open out completely enabling them to be secured instantly at the required position on any rod
  • Lock into place with a Labomeca® Allen key (optional, 241-0053)
* When 8 mm boss is locked onto the duralumin rod it cannot be moved by a rotational force less than 3 nm.
** When 8 to 12 mm boss is locked onto the duralumin rod it cannot be moved by a rotational force less than 6 nm.
*** When 16 mm boss is locked onto the duralumin rod it cannot be moved by a rotationalMore Product Information