Carboys, narrow neck, with screw cap
Supplier: KARTELL
1644 1646 1648 1650
216-2324EA 38.4 EUR
216-2324 216-2326 216-2328 216-2330
Carboys, narrow neck, with screw cap
Bottles Carboys
HDPE, translucent with blue screw cap.
  • Good chemical resistance
  • Heat resistant up to 80 °C (120 °C for short periods)
  • Heavy walled, very tough, a deeper closure with insert seal increasing leakproof performance
Ideal for storing and transporting liquids. Includes a carrying handle, 25 and 50 litre sizes have two handles.