Aspirator bottles with screw cap and spigot
Supplier: KARTELL
1660 1662 1664 1666
216-5745EA 57.5 EUR
216-5745 216-5744 216-5746 216-5747
Aspirator bottles with screw cap and spigot
Bottles Aspirators
HDPE, translucent with blue screw cap and tightly fitting spigot.
  • Good chemical resistance
  • Heat resistant up to 80 °C (120 °C for short periods)
  • Heavy walled, very tough, a deeper closure with insert seal increasing leakproof performance
Ideal for storing and dispensing liquids. Also recommended for use as settling bottles. Equipped with a carrying handle, the 25 and 50 litre sizes have two carrying handles.