Centrifuge bottles, PC/PPCO, with screw cap, Nalgene®
Supplier: Thermo Fisher Scientific
3140-0250 3140-0500 3141-0250 3141-0500
525-2472EA 160 EUR
525-2472 525-0047 525-2371 525-2373
Centrifuge bottles, PC/PPCO, with screw cap, Nalgene®
Bottles Centrifuge Bottles
PC or PPCO, wide-mouth, with PP screw closure with silicone gasket, autoclavable.
  • Excellent chemical resistance
  • Withstands forces up to 27 500×g for 250 ml and 13 700×g for 500 ml
  • When using the 250 ml bottle in Sorvall GSA rotor, use adapter 525-2300
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement