Dropping bottles, LDPE, Ranvier type
Supplier: KARTELL
388 389 391 392 393
215-9110EA 8.6 EUR
215-9110 215-9115 215-9120 215-9125 215-9130
Dropping bottles, LDPE, Ranvier type
Bottles Dropping Bottles
LDPE, translucent with red polyethylene screw cap.
  • Good chemical resistance
  • Heat resistance up to +80 °C
  • Leakproof
Bottles complete with dropping pipette and rubber teat.