Dropping bottles, FEP, Nalgene®
Supplier: Thermo Fisher Scientific
Nalgene®
215-7861EA 91.4 EUR
215-7861
Dropping bottles, FEP, Nalgene®
Bottles Dropping Bottles
PTFE/FEP, with ETFE drip lock and cap.
  • Excellent chemical resistance
  • Heat resistant up to 150 °C, autoclavable
  • Attached sealing cap cannot be lost
Suitable for drop-dispensing practically any chemical.