Jerricans, narrow neck
Supplier: RIXIUS
6-0203-200-61 6-0203-100-45 6-0203-250-61 6-0203-100-45B 6-0203-200-61B 6-0203-250-61B 6-0203-050-45B
216-5308EA 43 EUR
216-5308 216-5341 216-5342 RIXI6-0203-100-45B RIXI6-0203-200-61B RIXI6-0203-250-61B RIXI6-0203-050-45B
Jerricans, narrow neck
Bottles Jerricans
HDPE, blue.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
  • UN approval
Universal jerrican for industry, trade and laboratories. Ideal for packaging, storage and transportation.