Bottles, narrow neck, square, with screw caps, DURAN®
Supplier: DWK Life Sciences
218202453 218203655 218204454 218205459
215-8401EA 14.6 EUR
215-8401 215-8402 215-8403 215-8404
Bottles, narrow neck, square, with screw caps, DURAN®
Bottles Glass Bottles
DURAN®, borosilicate glass 3.3, clear, with PP screw cap and pouring ring.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Glass Type I according to USP, EP and JP
Graduated, DIN thread, pouring ring and blue screw cap. Save space on shelves, in the refrigerator, in autoclaves, or during transport. Stable, non-slip.
With printed retrace code (batch identification).