Bottles, narrow neck, round, HDPE, with screw caps, Boston type, Nalgene®
Supplier: Thermo Fisher Scientific
312002-9050 312002-0008 312002-9125 312002-0032 312002-0006 312002-0001 312002-0002 312002-0004 312002-0016 312002-9025
215-0234EA 1940 EUR
215-0234 215-0209 215-0246 215-0225 215-0207 215-0189 215-0190 215-0194 215-1377 215-1379
Bottles, narrow neck, round, HDPE, with screw caps, Boston type, Nalgene®
Bottles Plastic Bottles
HDPE, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant from –100 to +120 °C
  • Leakproof
Ideal for storing, packaging and transporting liquids. Robust, strong and stable.
Upozornení: The product(s) on this page are outside the scope of the EU Directive 2019/904.