Bottles, narrow neck, round, HDPE, with screw caps, sterile, Nalgene®
Supplier: Thermo Fisher Scientific
342002-9025 342002-9050 342089-0001 342089-0002 342089-0004 342089-0008 342089-0016 342089-0032
215-1163EA 981 EUR
215-1163 215-1164 215-1169 215-1170 215-1171 215-1173 215-1180 215-1181
Bottles, narrow neck, round, HDPE, with screw caps, sterile, Nalgene®
Bottles Plastic Bottles
HDPE, translucent with white PP screw cap.
  • Good chemical resistance
  • Heat resistant from −100 to +120 °C
  • Leakproof
Manufactured and packed under cleanroom conditions to minimise contamination with biological particles. Non toxic and non pyrogenic. Gamma sterilised. Packed in shrink-wrapped trays.
Compliant with the requirements of USP class VI.
Certifikace: Compliant with the requirements of USP class VI.