Bottles, narrow neck, round, LDPE, with screw caps, Azlon®
Supplier: SciLabware
BNL0060PN BNL0500P BNL1000P BNL0030PN BNL0150PN
215-0522EA 19.9 EUR
215-0522 215-0525 215-0526 215-4058 215-3108
Bottles, narrow neck, round, LDPE, with screw caps, Azlon®
Bottles Plastic Bottles
LDPE, translucent with PP screw cap.
  • Good chemical resistance, heat resistant up to +80 °C
  • More flexible and more impact resistant than HDPE bottles
Ideal for a wide range of liquid storage and sampling applications.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904