Screw caps
Supplier: GLASWARENFABRIK KARL HECHT
41326032 41326040 41326055 41326018 41326025 41326028
215-2803EA 0.4 EUR
215-2803 215-2802 HECH41326055 HECH41326018 HECH41326025 HECH41326028
Screw caps
Closures Bottle Closures
PP, black.
  • Very good chemical resistance
  • Heat-resistant up to +110°C
  • Only suitable for bottles 215-2814 to 215-2819