Wash bottles, narrow neck, Nalgene®
Supplier: Thermo Fisher Scientific
2403-0125 2403-0250 2403-0500 2403-1000
215-8121EA 163 EUR
215-8121 215-8122 215-8123 215-8124
Wash bottles, narrow neck, Nalgene®
Bottles Wash Bottles
PTFE/FEP, with ETFE spray cap.
  • Excellent chemical resistance
  • Heat resistant up to +205 °C, autoclavable
  • Leakproof
With excellent chemical resistance and low permeability. Ideal for contamination-free trace analysis work. Free of phthalates. Jet tip can be cut to increase flow rate.