Wash bottles, narrow neck
Supplier: Kautex
76082 76084
KAUT76082EA 3.6 EUR
KAUT76082 KAUT76084
Wash bottles, narrow neck
Bottles Wash Bottles
LDPE, translucent with LDPE spray cap.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Leakproof
Economical, soft and very easy to press. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The tip can be cut off to increase the flow rate.