VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
215-5680EA 81.5 EUR
215-5680 215-5679 215-5678 215-5677 215-5676
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
LDPE.
  • Amber coloured to protect light-sensitive substances
  • Leakproof, tough and versatile
Sample collection, storage and transport, for liquids, pastes and granular material. The wide mouth allows easy filling and cleaning.