VWR®, Bottles, Wide Neck, with Screw Cap
Supplier: VWR Collection
215-1152EA 32.2 EUR
215-1152 215-1153 215-1154 215-1155 215-1156 215-1157 215-1158 215-1159 215-1160 215-1161 215-1902
VWR®, Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
PTFE, strong walled.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls prevent contamination
Robust construction. Special recessed grips on the cap, and grooves in the flask, make it easy to handle. Non stick, the wide mouth allows easy filling and cleaning.