Laboratory and artist brushes
Supplier: VEREINIGTE PINSELFABRIKEN
659003 7700 7701 7702 7705 7706 SET-ROT 314G0 314G1 314G2 6200.5
149-2113EA 10.2 EUR
149-2113 149-2120 149-2121 149-2122 149-2125 149-2126 149-2127 149-2140 149-2141 149-2142 149-2153
Laboratory and artist brushes
Brushes
This brush range covers a wide variety of applications thanks to the number of styles on offer.
  • Paintbrush made of pale ox hair
  • Red marten brush — pure marten hair with black painted wooden handle
  • Goat-hair brush — oval, on wooden handle
  • Wide bear-hair brush, flat handle
Set 1: Contains 1 red marten brush in each size, sizes 1 to 5.