VWR® SafEPAk® Safety Bottle Containers
Supplier: VWR Chemicals
332493FEA 96.4 EUR
332493F 332497J
VWR® SafEPAk® Safety Bottle Containers
Carriers Bottle Carriers/Containers
These vessels contain chemical spillages, and being translucent, allow the examination of the condition and contents of the inner container without opening. Safepak® are designed to be used with BDH branded glass and Safebreak bottles.
HDPE.
  • Contains bottle spillages
  • For safe storage and transportation of chemicals
  • Good temperature resistance