Cell culture chambers, Nunc™ cell factories
Supplier: Thermo Fisher Scientific
139446 164327 167695 170009 140004 165250
734-2028EA 1510 EUR
734-2028 734-2082 732-2606 734-2105 734-2187 732-2603
Cell culture chambers, Nunc™ cell factories
Bioreactors
PS, sterile. For the industrial scale production of, for example, vaccines, monoclonal antibodies or pharmaceuticals.
  • Ideal for adherent cells, but can also be used for suspension cultures
  • Growth kinetics are unaltered from laboratory scale culture
  • Available in 1, 2, 10 and 40 tray versions for easy scale-up
  • Low contamination risk
  • Certified Nunclon™Δ surface treatment ensures excellent conditions for cell attachment and growth
Tray L×W: 335×205 mm.
Tray L×W: 335×205 mm
Informace o príslušenství: *Start-up kit includes: PC connector, air ventMore Product Information