Loop holders
Supplier: Copan
K44 K45 K46 K47
612-2683EA 38.6 EUR
612-2683 612-2684 612-2685 612-2686
Loop holders
Dissecting and Inoculating Loop and Needle Holders
Light weight aluminum or heavier nickel plated brass holders. The shafts of the loop holders are insulated with PVC. Both aluminum and brass holders are available in a choice of sizes 6" (150 mm) and 7" (175 mm).
Balení: Loop holders are purchased individually.