Burette clamps
Supplier: KARTELL
139 140
241-2020EA 48.5 EUR
241-2020 241-2021
Burette clamps
Clamps General Purpose Clamps
PP, white, robust. For fast and safe clamping of burettes.
  • Clamps fit rods 8 to 14 mm diameter
  • Clamps have rubber inserts and stainless steel springs
  • Burette scale not obstructed