VWR®, Clamps, Three Pronged, with Flexible Rod
Supplier: VWR
VWR®
241-0034EA 112 EUR
241-0034
VWR®, Clamps, Three Pronged, with Flexible Rod
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Jaws covered in vinyl to protect clamped items
  • Clamp rod diameter: 8 mm; rod length: 200 mm