VWR® Clamps, Three Pronged
Supplier: Avantor
VWR®
241-0019EA 251 EUR
241-0019
VWR® Clamps, Three Pronged
Clamps General Purpose Clamps
Laboral aluminium alloy.
  • Two easy action knurled screws for adjusting the clamping fingers independently of each other