Screw caps, PP, black with PE foam insert
Supplier: Kautex
805-70750 805-70751 805-70752 805-70753 805-70754 805-70755
215-3921EA 0.5 EUR
215-3921 215-3922 215-3923 215-3924 215-3925 215-3926
Screw caps, PP, black with PE foam insert
PP, black with PE foam insert.
  • Very good chemical resistance
  • Good temperature resistance
  • Leakproof
Suitable for wide mouth jars and containers.