Bottles, wide neck, with screw cap
Supplier: GRAVIS
BOO434 B00444B B00464 B00647 B00664 B00454 B00654
215-2602EA 92.3 EUR
215-2602 SPVAB00444B 215-3225 215-3227 215-3229 215-2799 215-3228
Bottles, wide neck, with screw cap
Bottles Glass Bottles
Glass, brown or clear.
  • Stoppers with a choice of 3 seals: TRISEAL (expanded PE foam coated on each side with PE film), and AS (laminated cork), (unbleached cardboard manufactured from wood fibre and coated on each side with PE film) or Butyl/PTFE
Informace pro objednávání: Bottles supplied without a stopper (to be ordered separately).