VWR®, Sleeve Protectors, LDPE/LLDPE
Supplier: VWR Collection
113-8256EA 22.3 EUR
113-8256 113-8257
VWR®, Sleeve Protectors, LDPE/LLDPE
Disposable Garments
Sleeve protectors made from 65% LDPE and 35% LLDPE.
  • Elasticated cuffs
  • Two colours available
Ø×L: 200×400 mm