HPLC columns, ZORBAX®
Supplier: Hichrom Limited
554-7541EA 488 EUR
HICHZNH-250A HICHZNH-150A
HPLC columns, ZORBAX®
ZORBAX® spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process. The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
  • Spherical porous silica
  • Narrow particles and pore size distribution
  • Robust with good batch to batch reproducibility
Informace pro objednávání: Other dimensions columns and guard cartridges are available on request.