HPLC columns, NUCLEOSIL®
Supplier: Hichrom Limited
HICHNC100-10SB-50AEA 392 EUR
HICHNC100-10SB-50A HICHNC100-10CN-50A
HPLC columns, NUCLEOSIL®
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 Å and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.
  • Good mechanical stability
  • Good batch to batch reproducibility