Coil condensers with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
05233314 05233211 05233414 05233514
201-1680EA 113 EUR
201-1680 201-0898 201-1075 201-1076
Coil condensers with standard ground joints
Condensers
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For cooling condensate from vapours, with cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.