VWR®, Jars, Wide Mouth, with Screw Cap
Supplier: VWR Collection
216-0164EA 48.8 EUR
216-0164 216-0165 216-0166 216-0167 216-0168 216-0169 216-0170 216-0171 216-0172
VWR®, Jars, Wide Mouth, with Screw Cap
Jars General Use Jars
PTFE, strong walled.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Stable, extremely robust construction. Special recessed grips in the cap make it easy to handle. The wide mouth allows easy filling and cleaning.