VWR®, Sample Containers, with Attached Lids
Supplier: VWR Collection
216-2625EA 220 EUR
216-2625 216-2627 216-2628
VWR®, Sample Containers, with Attached Lids
Sample Containers General Purpose Containers
PP, translucent.
  • Can be used with one hand; the cap remains vertical when open
  • Very good chemical and temperature resistance
  • Aseptic
Ideal for collecting, storing and transporting samples.
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904.