VWR® Hard Bottle Pack
Supplier: Avantor
VWR®
216-1168EA 225 EUR
216-1168 216-1169
VWR® Hard Bottle Pack
Cooling Packs
These tough, resuable bottle packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator and are effective when used with insulated shipping containers.
  • Hard plastic bottle pack is filled with water
  • Can be refrigerated or frozen to keep products cold or frozen
  • Long lasting, high-performance temperature protection
These robust ice packs are suitable for cooling products in cold boxes, pouches or packages. The gel ensures constant and reliable cooling.
  • Non toxic for use with foodstuffs
  • Suitable for heavy loads
  • Adapted to rough transport conditions
Hard bottle packs are suitable for use with pharmaceuticals, clinical trials, diagnostic kits, chemicals, and biologicals. Phase changes at 0 °C and returns to liquid state after thawing.