Desiccant canister, SP Bel-Art Desi-Can™
Supplier: Bel-Art Products, a part of SP
Desi-Can™
467-0000EA 49.4 EUR
467-0000
Desiccant canister, SP Bel-Art Desi-Can™
Desiccators Desiccants
Reusable canister which is ideal for use in desiccator cabinets, cupboards, containers, tool boxes and other storage areas to prevent rust and moisture. The silica gel inside effectively absorbs moisture through perforations in the aluminium case.
  • One canister will maintain a safe level of humidity in 0,9 m³ (3 ft³)
  • Can be regenerated in an oven at 150 °C for about 3 hours giving unlimited life
  • Monitor window shows blue when desiccant is recharged and pink when saturated