VWR®, Dishes, Evaporating, Tall Form
Supplier: VWR
VWR®
216-0183EA 44 EUR
216-0183 216-0184 216-0181 216-0182 216-0185 216-0179 216-0180
VWR®, Dishes, Evaporating, Tall Form
Trays/Dishes/Pots Evaporating Basins/Dishes
PTFE, with spout.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth inner walls to prevent contamination
Thick walled, conical shape. Extremely robust.