VWR®, Dishes, Evaporating
Supplier: VWR Collection
216-0083EA 9.5 EUR
216-0083 216-0084 216-0085 216-0086 216-0087 216-0088 216-0089 216-0090 216-0091 216-0092
VWR®, Dishes, Evaporating
Trays/Dishes/Pots Evaporating Basins/Dishes
Borosilicate 3.3 glass, with spout.
  • Very good chemical resistance
  • High temperature resistance up to +500 °C
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
The economical alternative for a wide range of applications. With flat bottom and white labelling area.
DIN 12336
Certifikace: DIN 12336