Petri dish dispenser
Supplier: Heathrow Scientific
391-1165EA 116 EUR
391-1165
Petri dish dispenser
Racks Petri Dish Racks
Acrylic. This sturdy dispenser will hold up to twenty Ø 90 mm Petri dishes in two easy to access partitions.
  • Mounting screws included
  • On the counter or on a wall
  • Solid and clear design conception
W×D×H: 203×102×229 mm