VWR®, Disposable Weighing Dishes with Tabs
Supplier: VWR
VWR®
611-1379EA 32.1 EUR
VWRU12577-081 VWRU16005-126_U 611-1379 611-1378 VWRU16005-126 611-1362 611-1372 611-1371
VWR®, Disposable Weighing Dishes with Tabs
Boats Weighing Boats
Aluminium.
  • Flat bottom and crimped sides for rigidity
  • Dishes feature tabs for easy handling
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes feature crimped sides and tabs. Ideal for general purpose weighing, dispensing and storage applications. Can also serve as evaporating dishes, parts containers or dust covers.
  • Flat bottom and crimped sides for rigidity
  • Dishes feature tabs for easy handling
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes feature crimped sides and tabs. Ideal for general purpose weighing, dispensing and storage applications. Can also serve as evaporating dishes, parts containers or dust covers.