Disposable earplugs, Laser Lite®
Supplier: HONEYWELL SAFETY
3301105 3301106 1013047
111-0137EA 49.9 EUR
111-0137 111-0138 111-0395
Disposable earplugs, Laser Lite®
Earplugs Disposable Earplugs
Laser Lite® earplugs offer optimal comfort for long-term wear.
  • Bright colours for high visibility
  • Self-adjusting foam to fit virtually every user
  • T-shape for easy insertion and wear
SNR: 35 dB
EN 352-2
Certifikace: EN 352-2
Upozornení: * One size fits all.