Reusable earplugs, Fusion™
Supplier: HONEYWELL SAFETY
1011282 1011281
111-0115EA 159 EUR
111-0115 111-0084
Reusable earplugs, Fusion™
Earplugs Reusable Earplugs
Fusion™ earplugs provide a combination of all-day comfort and easy handling.
  • Patented dual material design
  • Two sizes provide optimal fit and safety
  • Unique detachable cord system
SNR: 28 dB
EN 352-2
Certifikace: EN 352-2