Banded earplugs, PerCap®
Supplier: Honeywell Safety
PerCap®
111-3581EA 6.9 EUR
111-3581
Banded earplugs, PerCap®
Earplugs Banded Earplugs
This foldable headband with soft foam plugs offers a comfortable fit.
  • Perfect solution for temporary need for hearing protection
  • Lightweight and foldable
  • Earplugs easily fit into a pocket when not needed
SNR: 24 dB
Certifikace: EN 352-2
Delivery information: Supplied with a storage case.