Groundwater sampling capsules, Aquaprep™ 600
Supplier: Cytiva (Formerly Pall Lab)
12175 12176
300-0005EA 58.8 EUR
300-0005 516-7531
Groundwater sampling capsules, Aquaprep™ 600
Filters Membrane Filter Capsules
Aquaprep™ 600 capsules are designed for the preparation of groundwater samples for dissolved metals analysis. 42 metals Analysis Certification is printed on each package.
  • Meets filtration requirements of the U.S. EPA. for 0,45 µm filtration
  • Reduces the need for multiple filter changes when filtering silt and particulate laden groundwater and ensures rapid filtration
  • Aquaprep™ 600 has four times the filtration area of conventional 142 mm filters
Supor® membrane (hydrophilic polyethersulphone) in a PP housing. Effective filtration area: 600 cm²; maximum pressure 4,1 bar.
Dimensions: Ø 68 mm, length 145 mm
Input/output: AquaPrep™ device: Stepped hose barb accepts 6,4 to 12,7 mm Int.Ø tubing.