Venting filters, Midisart® 2000
Supplier: Sartorius
17805-TCN
517-0014EA 0 EUR
517-0014
Venting filters, Midisart® 2000
Filters Venting Filters
Venting filters for the sterile venting of small fermenters and culture vessels.
Hydrophobic PP reinforced PTFE membrane, PP housing
Order Now
SPECIFICATIONS
Pore size 0,2 µm
Environmentally Preferable
Airflow rate 5 L/min (0,1 bar)
Max. pressure 3 bar
Max. temperature 134 °C