First aid kit
Supplier: Söhngen
118-1410EA 886 EUR
118-1410
First aid kit
First Aid Supplies First Aid Kits
This customised, professional first aid kit is suitable for use in laboratories and the chemical industry. It contains an eyewash fountain, cold compress, skin disinfectant, dressings, etc.
  • Orange ABS case
  • Wall mounting is possible
W×D×H: 400×300×150 mm
Certifikace: DIN 13157