Erlenmeyer flasks, wide neck, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
4108-0125 4108-0250 4108-0500
214-7202EA 110 EUR
214-7202 214-7203 214-7204
Erlenmeyer flasks, wide neck, with screw caps, Nalgene®
Flasks Erlenmeyer Flasks
PC, transparent with PP screw cap.
  • Robust, safer than glass flasks when used with shakers
  • Flasks and caps can be autoclaved However, repeated autoclaving can impair both the mechanical strength and the visual clarity of the flasks
  • Linerless tight sealing closure protecting contens from spills and contamination
For preparing and storing nutrient media and for many culturing procedures.