Fernbach culture flasks, Nalgene®
Supplier: Thermo Fisher Scientific
Nalgene®
710-4100EA 73.75 EUR
710-4100
Fernbach culture flasks, Nalgene®
Flasks Culture Flasks
Clear PC.
  • Thermal resistance from -135 to +135 °C, autoclavable
  • Shatter-proof with high mechanical resistance
  • Good chemical resistance, but not suitable for solvents, strong acids or alkaline solutions
Provides the large liquid surface area necessary for growing microorganisms in liquid culture media. Provides for faster growth through  improved ventilation.