Flasks, flat bottom with standard ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
03011928 03011937 03011949 03011958 03012428 03012437 03012449 03012458 03012470 03012473 03012928 03012937 03012949 03012958 03012970 03012973
201-1211EA 13.7 EUR
201-1211 201-1212 201-1213 201-1214 201-1311 201-1312 201-1313 201-1314 201-1315 201-1316 201-1201 201-1202 201-1203 201-1204 271-1205 271-1206
Flasks, flat bottom with standard ground joint
Flasks Flat Bottom Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
DIN 12348
Certifikace: DIN 12348