Flasks, pear shaped with standard ground joint
Supplier: Lenz Laborglas GmbH & CO.KG
03031413 03031422 03031428 03031437 03031922 03031928 03031937 03031949 03031449 03031410 03032928 03032937 03032949
201-1221EA 9.9 EUR
201-1221 201-1222 201-1223 201-1224 201-1041 201-0321 201-0322 201-1042 201-1040 201-0320 201-1043 201-1044 201-1045
Flasks, pear shaped with standard ground joint
Flasks Pear Shaped Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint socket and white labelling area.