Volumetric flasks, class B, white markings, SILBERBRAND
Supplier: Brand
36753 36754 36747 36751 36752
612-5060EA 0 EUR
612-5060 612-5061 612-5054 612-5058 612-5059
Volumetric flasks, class B, white markings, SILBERBRAND
Flasks Volumetric Flasks
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast white enamel
*Wide neck.
Certifikace: DIN EN ISO 1042